Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TIM-3 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2365C
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2365C 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2365C Supplier R&D Systems Supplier No. FAB2365C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TIM-3
Applications Flow Cytometry
Classification Monoclonal
Clone 344823
Conjugate PerCP
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8TDQ0.2
Gene Alias CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Gene Symbols HAVCR2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIM-3 Ser22-Arg200 Accession # Q8TDQ0.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84868
Test Specificity Detects human TIM-3 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) TIM-1, rhTIM-4, recombinant mouse (rm) TIM-1, rmTIM-2, rmTIM-3, rmTIM-5, or rmTIM-6.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.