Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TIM-3 Quantikine ELISA Kit

Catalog No. dtim30
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

For the quantitative determination of human T cell Immunoglobulin and Mucin domain 3 (TIM-3) concentrations in cell culture supernates, cell lysates, serum, plasma, and urine. Human TIM-3 Immunoassay employs the quantitative sandwich enzyme immunoassay technique.

The Human TIM-3 Quantikine ELISA Kit quantifies human TIM-3 in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (10 ul).

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 8.75 pg/mL
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human TIM-3
For Use With (Application) ELISA
Gene ID (Entrez) 84868
Gene Symbol HAVCR2
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Target TIM-3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.