Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TIM-4 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2929T025
This item is not returnable.
View return policy
Description
TIM-4 Monoclonal specifically detects TIM-4 in Human samples. It is validated for Flow Cytometry.Specifications
TIM-4 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD4, TIMD-4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
91937 | |
Human | |
Purified |
Flow Cytometry | |
921832 | |
Flow Cytometry 5 uL/10^6 cells | |
Q96H15 | |
TIMD4 | |
Mouse myeloma cell line NS0-derived recombinant human TIM-4 Glu25-Leu315 Accession # Q96H15 | |
25 Tests | |
Primary | |
Detects human TIM-4 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction