Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TIM-4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2929
This item is not returnable.
View return policy
Description
TIM-4 Polyclonal specifically detects TIM-4 in Human samples. It is validated for Western Blot.Specifications
TIM-4 | |
Polyclonal | |
Biotin | |
Q96H15 | |
TIMD4 | |
Mouse myeloma cell line NS0-derived recombinant human TIM-4 Glu25-Leu315 Accession # Q96H15 | |
50 μg | |
Primary | |
Detects human TIM-4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) TIM-1, recombinant mouse (rm) TIM-2, rhTIM-3, rmTIM-4, rmTIM-5, and rmTIM-6 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD4, TIMD-4 | |
Goat | |
Affinity Purified | |
RUO | |
91937 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction