Learn More
Human TIMP-1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | TIMP-1 |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA |
| Classification | Monoclonal |
| Clone | 63515 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P01033 |
| Gene Alias | CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 |
| Gene Symbols | TIMP1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.