Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TIMP-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF970
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF970 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF970 Supplier R&D Systems Supplier No. AF970
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 13 publications

TIMP-1 Polyclonal specifically detects TIMP-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization, Knockout Validated.

Specifications

Antigen TIMP-1
Applications Western Blot, Immunohistochemistry, Neutralization, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 1-15 ug/mL, Neutralization 1 ug/mL, Knockout Validated
Gene Accession No. Q6FGX5
Gene Alias CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Gene Symbols TIMP1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIMP-1 Cys24-Ala207 Accession # Q6FGX5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7076
Test Specificity Detects human TIMP-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse TIMP-1 and recombinant rat TIMP-1 is observed and less than 1% cross-reactivity with recombinant human (rh) TIMP-2, rhTIMP-3, and rhTIMP-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.