Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TIMP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $1885.00
Specifications
Antigen | TIMP-2 |
---|---|
Clone | 127711 |
Dilution | Knockout Validated 4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Immunoassay, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9711SP
![]() |
R&D Systems
MAB9711SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9711100
![]() |
R&D Systems
MAB9711100 |
100 μg |
Each for $514.50
|
|
|||||
MAB9711500
![]() |
R&D Systems
MAB9711500 |
500 μg |
Each for $1,885.00
|
|
|||||
Description
TIMP-2 Monoclonal specifically detects TIMP-2 in Human samples. It is validated for Knockout Validated, ELISA Capture (Matched Antibody Pair).Specifications
TIMP-2 | |
Knockout Validated 4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P16035 | |
7077 | |
Chinese hamster ovary cell line CHO-derived recombinant human TIMP-2 Cys27-Pro220 Accession # P16035 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TIMP-2 in ELISAs. |
127711 | |
Immunoassay, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
TIMP2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title