Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TIMP-3 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC9731V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9731V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9731V100 Supplier R&D Systems Supplier No. IC9731V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TIMP-3 Monoclonal specifically detects TIMP-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TIMP-3
Applications Flow Cytometry
Classification Monoclonal
Clone 277128
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P35625
Gene Alias HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3
Gene Symbols TIMP3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIMP-3 Cys24-Pro211 Accession # P35625
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7078
Test Specificity Detects human TIMP-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TIMP-1, -2, or -4 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.