Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TINAGL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7185SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7185SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7185SP Supplier R&D Systems Supplier No. AF7185SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TINAGL1 Polyclonal specifically detects TINAGL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TINAGL1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9GZM7
Gene Alias androgen-regulated gene 1, ARG1, GIS5, Glucocorticoid-inducible protein 5, LCN7, LIECG3, lipocalin 7, OLRG2, OLRG-2, Oxidized LDL-responsive gene 2 protein, oxidized-LDL responsive gene 2, P3ECSL, TIN Ag-related protein, TINAGL, TINAG-like 1, TIN-Ag-RP, TINAGRP, tubulointerstitial nephritis antigen-like, tubulointerstitial nephritis antigen-like 1, Tubulointerstitial nephritis antigen-related protein
Gene Symbols TINAGL1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TINAGL1 Ala22-Met464 Accession # Q9GZM7
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64129
Test Specificity Detects human TINAGL1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TINAG is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.