Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TINAGL1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7185
This item is not returnable.
View return policy
Description
TINAGL1 Polyclonal specifically detects TINAGL1 in Human samples. It is validated for Western Blot.Specifications
TINAGL1 | |
Polyclonal | |
Unconjugated | |
Q9GZM7 | |
TINAGL1 | |
Chinese hamster ovary cell line CHO-derived recombinant human TINAGL1 Ala22-Met464 Accession # Q9GZM7 | |
100 μg | |
Primary | |
Detects human TINAGL1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TINAG is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.25 ug/mL | |
androgen-regulated gene 1, ARG1, GIS5, Glucocorticoid-inducible protein 5, LCN7, LIECG3, lipocalin 7, OLRG2, OLRG-2, Oxidized LDL-responsive gene 2 protein, oxidized-LDL responsive gene 2, P3ECSL, TIN Ag-related protein, TINAGL, TINAG-like 1, TIN-Ag-RP, TINAGRP, tubulointerstitial nephritis antigen-like, tubulointerstitial nephritis antigen-like 1, Tubulointerstitial nephritis antigen-related protein | |
Sheep | |
Affinity Purified | |
RUO | |
64129 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction