Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tissue alpha-L-Fucosidase/FUCA1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7039SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7039SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7039SP Supplier R&D Systems Supplier No. MAB7039SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tissue alpha-L-Fucosidase/FUCA1 Monoclonal specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Tissue alpha-L-Fucosidase/FUCA1
Applications Western Blot
Classification Monoclonal
Clone 810607
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P04066
Gene Alias Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, FUCA1, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase, Tissue alphaLFucosidase
Gene Symbols FUCA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tissue alpha-L-Fucosidase/FUCA1 Gln32-Lys466 Accession # P04066
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2517
Test Specificity Detects human Tissue alpha-L-Fucosidase/FUCA1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.