Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TJAP1 Partial ORF (NP_542171.1, 77 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964817
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a tight junction-associated protein. Incorporation of the encoded protein into tight junctions occurs at a late stage of formation of the junctions. The encoded protein localizes to the Golgi and may function in vesicle trafficking. Alternatively spliced transcript variants have been described. A related pseudogene exists on the X chromosome. [provided by RefSeq]

Sequence: LELELGQSREELDKFKDKFRRLQNSYTASQRTNQELEDKLHTLIKKAEMDRKTLDWEIVELTNKLLDAKNTINKLEELNERYRLDCNPAVQLLKCNKSHFRNHK

Specifications

Accession Number NP_542171.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 93643
Molecular Weight (g/mol) 37.18kDa
Name TJAP1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LELELGQSREELDKFKDKFRRLQNSYTASQRTNQELEDKLHTLIKKAEMDRKTLDWEIVELTNKLLDAKNTINKLEELNERYRLDCNPAVQLLKCNKSHFRNHK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686F06131/PILT/TJP4
Common Name TJAP1
Gene Symbol TJAP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.