Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TL-1A, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF31023100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF31023100 100 μg
AF310231MG 1 mg
AF3102320UG 20 μg
AF31023500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF31023100 Supplier Gibco™ Supplier No. AF31023100UG
  • $772.00
    $579.65 / Each of 1

    Save $192.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TL-1A is a 22.1 kDa polypeptide of 195 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein is abundantly expressed in endothelial cells, but is not expressed in either B or T cells. The expression of this protein is inducible by TNF and IL-1 alpha. This cytokine is a ligand for receptor TNFRSF25 and decoy receptor TNFRSF21/DR6. It can activate NF-kappaB and MAP kinases, and acts as an autocrine factor to induce apoptosis in endothelial cells. This cytokine is also found to inhibit endothelial cell proliferation, and thus may function as an angiogenesis inhibitor. An additional isoform encoded by an alternatively spliced transcript variant has been reported but the sequence of this transcript has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95150
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 9966
Molecular Weight (g/mol) 22.1 kDa
Name Human TL-1A, Animal-Free
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias bM20K13.3 (tumor necrosis factor (ligand) superfamily, member 15); MGC129934; MGC129935; TL1; TL1A; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; TNFSF15; TNLG1B; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; tumor necrosis factor superfamily member 15; vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor; vascular endothelial growth inhibitor-192A; Vegi; VEGI192A
Common Name TL1A
Gene Symbol TNFSF15
Biological Activity Determined by its ability to stimulate IFN-gamma production by human PBMC co-stimulated with Human IL-12 and Human IL-18.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQLRAQGEAS VQFQALKGQE FAPSHQQVYA PLRADGDKPR AHLTVVRQTP TQHFKNQFPA LHWEHELGLA FTKNRMNYTN KFLLIPESGD YFIYSQVTFR GMTSECSEIR QAGRPNKPDS ITVVITKVTD SYPEPTQLLM GTKSVCEVGS NWFQPIYLGA MFSLQEGDKL MVNVSDISLV DYTKEDKTFF GAFLL
Content And Storage -20°C
Activity Determined by its ability to stimulate IFN-gamma production by human PBMC co-stimulated with Human IL-12 and Human IL-18.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.