Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TL1A/TNFSF15 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB74422U100UG
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Flow Cytometry.Specifications
TL1A/TNFSF15 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1 | |
E. coli-derived recombinant human TL1A/TNFSF15, Leu72-Leu251, Accession No. O95150 | |
100 μg | |
9966.0 | |
Human | |
Supernatant |
Flow Cytometry | |
2116A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O95150 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human TL1A/TNFSF15 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction