Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLE1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5947
This item is not returnable.
View return policy
Description
TLE1 Polyclonal specifically detects TLE1 in Human samples. It is validated for Western Blot.Specifications
TLE1 | |
Polyclonal | |
Unconjugated | |
Q04724 | |
TLE1 | |
E. coli-derived recombinant human TLE1 His11-Asn200 Accession # Q04724 | |
100 μg | |
Primary | |
Detects human TLE1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) TLE3 and rhTLE4 is observed and less than 5% cross-reactivity with rhTLE2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
Sheep | |
Affinity Purified | |
RUO | |
7088 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction