Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLE2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7216
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7216 Supplier R&D Systems Supplier No. AF7216
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TLE2 Polyclonal specifically detects TLE2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen TLE2
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q04725
Gene Alias enhancer of split groucho 2, ESG, ESG2, ESG2Enhancer of split groucho-like protein 2, FLJ41188, GRG2, transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 2, homolog of Drosophila E(sp1), transducin-like enhancer protein 2
Gene Symbols TLE2
Host Species Sheep
Immunogen E. coli-derived recombinant human TLE2 Ile11-Ser193 Accession # Q04725
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7089
Test Specificity Detects human TLE2 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) TLE1, rhTLE3, and rhTLE4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.