Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR1 APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB1484A
This item is not returnable.
View return policy
Description
TLR1 Polyclonal specifically detects TLR1 in Human samples. It is validated for Flow Cytometry.Specifications
TLR1 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1 | |
Goat | |
Affinity Purified | |
RUO | |
7096 | |
Human | |
IgG |
Flow Cytometry | |
APC | |
AAC34137 | |
TLR1 | |
Mouse myeloma cell line NS0-derived recombinant human TLR1 Ser22-Asn578 Accession # AAC34137 | |
100 Tests | |
Primary | |
Detects human TLR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) TLR1 is observed and less than 5% cross-reactivity with recombinant human (rh) TLR3, rhTLR4, and rmTLR6 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction