Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1484
This item is not returnable.
View return policy
Description
TLR1 Polyclonal specifically detects TLR1 in Human samples. It is validated for Western Blot.Specifications
TLR1 | |
Polyclonal | |
Biotin | |
Q15399 | |
TLR1 | |
Mouse myeloma cell line NS0-derived recombinant human TLR1 Ser22-Asn578 (Asn228Ser) Accession # Q15399 | |
50 μg | |
Primary | |
Detects human TLR1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) TLR1 and rmTLR6 is observed and less than 1% cross-reactivity with recombinant human (rh) TLR3 and rhTLR4 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1 | |
Goat | |
Affinity Purified | |
RUO | |
7096 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction