Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLR10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6619
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6619 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6619 Supplier R&D Systems Supplier No. MAB6619
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Western Blot.

Specifications

Antigen TLR10
Applications Western Blot
Classification Monoclonal
Clone 670719
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BXR5
Gene Alias CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10
Gene Symbols TLR10
Host Species Mouse
Immunogen E. coli-derived recombinant human TLR10 Asn168-Lys383 Accession # Q9BXR5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81793
Test Specificity Detects human TLR10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TLR1, 2, 3, 4, 5, 7, 8, 9, recombinant mouse (rm) TLR6, 11, 12, rhIL-1 RI, rhIL-1 RII, rhIL-1 RAcP, rhST2, rhIL-18 R, rhIL-1 Rrp2, rhIL-18 R beta , rhSIGIRR, rhIL-1 RAPL1, rhTIGIRR, rhMD-1, rhMD-2, rhTIRAP, or rmRP105 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.