Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF2616SP
This item is not returnable.
View return policy
Description
TLR2 Polyclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
TLR2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD282, CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
Goat | |
Affinity Purified | |
RUO | |
7097 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF, ELISA | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
O60603 | |
TLR2 | |
Mouse myeloma cell line NS0-derived recombinant human TLR2 Glu21-Leu590 Accession # O60603 | |
25 μg | |
Primary | |
Detects human TLR2 in ELISAs and Western blots. In sandwich ELISAs, less than 1% cross-reactivity with recombinant mouse TLR2 and less than 0.3% cross-reactivity with recombinant human (rh) TLR3 and rhTLR4/MD-2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction