Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2616
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2616 Supplier R&D Systems Supplier No. AF2616
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

TLR2 Polyclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TLR2
Applications Western Blot, Flow Cytometry, CyTOF, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. O60603
Gene Alias CD282, CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Gene Symbols TLR2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TLR2 Glu21-Leu590 Accession # O60603
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7097
Test Specificity Detects human TLR2 in ELISAs and Western blots. In sandwich ELISAs, less than 1% cross-reactivity with recombinant mouse TLR2 and less than 0.3% cross-reactivity with recombinant human (rh) TLR3 and rhTLR4/MD-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.