Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human TLR3 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Description
The Human TLR3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TLR3.
Specifications
Specifications
Assay Range | 93.80 to 6000 pg/mL |
Product Type | Solid Phase Sandwich ELISA |
Gene ID (Entrez) | 7098 |
Gene Symbol | TLR3 |
Host Species | Human |
Quantity | 1 Kit (5 Plates) |
Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction