Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 17 publications
Supplier: R&D Systems AF1478SP
This item is not returnable.
View return policy
Description
TLR4 Polyclonal specifically detects TLR4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready.Specifications
TLR4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Goat | |
Affinity Purified | |
RUO | |
7099 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
LYOPH | |
Flow Cytometry 2.5 μg/10^6 cells, Immunohistochemistry 5-15 μg/mL, Neutralization 1.5-7.5 μg/mL, Immunocytochemistry 5-15 μg/mL, CyTOF-ready | |
O00206 | |
TLR4 | |
Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206 | |
25 μg | |
Primary | |
Detects human TLR4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) TLR4 is observed and less than 5% cross-reactivity with recombinant human (rh) TLR1, rhTLR2, rhTLR3 and rmTLR6 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction