Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB1478
This item is not returnable.
View return policy
Description
TLR4 Monoclonal specifically detects TLR4 in Human samples. It is validated for Western Blot.Specifications
TLR4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O00206 | |
TLR4 | |
Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206 | |
100 μg | |
Primary | |
Detects human TLR4 in direct ELISAs and Western blots. Shows approximately 25% cross-reactivity with recombinant mouse (rm) TLR4 and no cross-reactivity with recombinant human TLR1, 2, 3, or rmTLR6. | |
Human | |
Purified |
Western Blot | |
285227 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7099 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction