Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLR4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6248SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6248SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6248SP Supplier R&D Systems Supplier No. MAB6248SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR4 Monoclonal specifically detects TLR4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen TLR4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 610015
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00206
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7099
Test Specificity Detects human TLR4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TLR1, 2, 3, 5, 7, 8, 9, 10, recombinant mouse (rm) TLR4, rmTLR6, rhTIRAP, rhIL-1 RI, rhIL-1 RII, rhIL-1 RAcP, rhST2, rhIL-18 R, rhIL-1 Rrp2, rhIL-18 R beta , rhSIGIRR, rhIL-1 RAPL1, rhIL-1 RAPL2, rhMD-1, rhMD-2, or rmRP105 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.