Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLR4 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6248P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6248P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6248P Supplier R&D Systems Supplier No. FAB6248P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

TLR4 Monoclonal specifically detects TLR4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TLR4
Applications Flow Cytometry
Classification Monoclonal
Clone 610015
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O00206
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7099
Test Specificity Detects human TLR4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TLR1, 2, 3, 5, 7, 8, 9, 10, recombinant mouse (rm) TLR4, rmTLR6, rhTIRAP, rhIL-1 RI, rhIL-1 RII, rhIL-1 RAcP, rhST2, rhIL-18 R, rhIL-1 Rrp2, rhIL-18 R beta , rhSIGIRR, rhIL-1 RAPL1, rhIL-1 RAPL2, rhMD-1, rhMD-2, or rmRP105 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.