Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR5 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6704U100UG
This item is not returnable.
View return policy
Description
TLR5 Monoclonal specifically detects TLR5 in Human samples. It is validated for Flow Cytometry.Specifications
| TLR5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O60602 | |
| TLR5 | |
| E. coli-derived recombinant human TLR5 Ile21-Phe115 Accession # O60602 | |
| RUO | |
| 7100 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 624915 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MGC126430, MGC126431, SLEB1FLJ10052, TIL-3, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TLR5 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) TLR1, 2, 3, 4, 7, 8, recombinant mouse TLR5, 6, or rhTIRAP is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction