Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLR5 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6704G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6704G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6704G Supplier R&D Systems Supplier No. FAB6704G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR5 Monoclonal specifically detects TLR5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TLR5
Applications Flow Cytometry
Classification Monoclonal
Clone 624915
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O60602
Gene Alias MGC126430, MGC126431, SLEB1FLJ10052, TIL-3, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
Gene Symbols TLR5
Host Species Mouse
Immunogen E. coli-derived recombinant human TLR5 Ile21-Phe115 Accession # O60602
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7100
Test Specificity Detects human TLR5 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) TLR1, 2, 3, 4, 7, 8, recombinant mouse TLR5, 6, or rhTIRAP is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.