Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR7 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5875S100UG
This item is not returnable.
View return policy
Description
TLR7 Monoclonal specifically detects TLR7 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TLR7 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NYK1 | |
TLR7 | |
E. coli-derived recombinant human TLR7 Met360-Leu516 Accession # Q9NYK1 | |
RUO | |
51284 | |
Human | |
IgG2a |
Flow Cytometry | |
533707 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
toll-like receptor 7 | |
Mouse | |
100 μg | |
Primary | |
Detects human TLR7 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction