Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB5875SP
This item is not returnable.
View return policy
Description
TLR7 Monoclonal specifically detects TLR7 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
TLR7 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q9NYK1 | |
TLR7 | |
E. coli-derived recombinant human TLR7 Met360-Leu516 Accession # Q9NYK1 | |
25 μg | |
Primary | |
Detects human TLR7 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
533707 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
toll-like receptor 7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51284 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction