Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR8 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC8999G100UG
This item is not returnable.
View return policy
Description
TLR8 Monoclonal specifically detects TLR8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TLR8 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
Chinese hamster ovary cell line CHO-derived recombinant human TLR8 Glu27-Thr827 Accession # Q9NR97 | |
RUO | |
51311 | |
Human | |
IgG2b |
Flow Cytometry | |
935166 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NR97 | |
Mouse | |
100 μg | |
Primary | |
Detects human TLR8 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction