Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TLR8 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC8999N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8999N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC8999N100 Supplier R&D Systems Supplier No. IC8999N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR8 Monoclonal specifically detects TLR8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TLR8
Applications Flow Cytometry
Classification Monoclonal
Clone 935166
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9NR97
Gene Alias CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TLR8 Glu27-Thr827 Accession # Q9NR97
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51311
Test Specificity Detects human TLR8 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.