Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR9 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC36582G100UG
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TLR9 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD289, CD289 antigen, toll-like receptor 9 | |
Human TLR9 synthetic peptide Accession # Q9NR96 | |
100 μg | |
54106 | |
Human | |
Supernatant |
Flow Cytometry | |
980914 | |
Intracellular Staining by Flow Cytometry 0.25-1μg/106 cells | |
Q9NR96 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human TLR9 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction