Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $549.50
Specifications
| Antigen | TLR9 |
|---|---|
| Clone | 26C593R |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB36583SP
![]() |
R&D Systems
MAB36583SP |
25 μg |
Each for $214.00
|
|
|||||
MAB36583100
![]() |
R&D Systems
MAB36583100 |
100 μg |
Each for $549.50
|
|
|||||
Description
TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| TLR9 | |
| Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Antibody developed against the KLH-conjugated synthetic peptide corresponding to amino acids Asp250-Asn300 of human TLR9 isoform A. Accession # Q9NR96 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TLR9 in direct ELISAs. |
| 26C593R | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Human | |
| Q9NR96 | |
| 54106 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title