Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TM4SF1/L6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8164
This item is not returnable.
View return policy
Description
TM4SF1/L6 Monoclonal specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
TM4SF1/L6 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
L6, M3s1, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human TM4SF1/L6 in ELISA. In Flow Cytometry, it stains HEK293 cells transfected with human TM4SF1/L6, and does not stain HEK293 cells transfected with irrelevant human cell surface marker. | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
877621 | |
Unconjugated | |
P30408 | |
TM4SF1 | |
NS0 mouse myeloma cell line transfected with human TM4SF1/L6 Met1-Cys202 Accession # P30408 | |
100 μg | |
4071 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction