Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TM4SF4 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7998V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7998V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7998V100 Supplier R&D Systems Supplier No. FAB7998V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TM4SF4 Monoclonal specifically detects TM4SF4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TM4SF4
Applications Flow Cytometry
Classification Monoclonal
Clone 832441
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P48230
Gene Alias FLJ31015, il-TMP, ILTMP, ILTMPintestinal and liver (il) tetraspan membrane protein, Intestine and liver tetraspan membrane protein, transmembrane 4 L six family member 4, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4
Gene Symbols TM4SF4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TM4SF4 Accession # P48230
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7104
Test Specificity Detects human TM4SF4 in ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.