Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TM4SF4 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7998G100UG
This item is not returnable.
View return policy
Description
TM4SF4 Monoclonal specifically detects TM4SF4 in Human samples. It is validated for Flow Cytometry.Specifications
TM4SF4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P48230 | |
TM4SF4 | |
NS0 mouse myeloma cell line transfected with human TM4SF4 Accession # P48230 | |
RUO | |
7104 | |
Human | |
IgG1 |
Flow Cytometry | |
832441 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ31015, il-TMP, ILTMP, ILTMPintestinal and liver (il) tetraspan membrane protein, Intestine and liver tetraspan membrane protein, transmembrane 4 L six family member 4, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4 | |
Mouse | |
100 μg | |
Primary | |
Detects human TM4SF4 in ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction