Learn More
Human TM4SF4 APC-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | TM4SF4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 832441 |
| Conjugate | APC |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P48230 |
| Gene Alias | FLJ31015, il-TMP, ILTMP, ILTMPintestinal and liver (il) tetraspan membrane protein, Intestine and liver tetraspan membrane protein, transmembrane 4 L six family member 4, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4 |
| Gene Symbols | TM4SF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.