Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TMED1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2243T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2243T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2243T100 Supplier R&D Systems Supplier No. FAB2243T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMED1 Monoclonal specifically detects TMED1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TMED1
Applications Flow Cytometry
Classification Monoclonal
Clone 1009527
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q13445
Gene Alias IL1RL1-Binding Protein, Il1rl1l, IL1RL1LG, IL-1RL1LG, IL1RL1LGIL1RL1-binding protein, Interleukin 1 Receptor-Like 1 Ligand, Interleukin-1 receptor-like 1 ligand, Ly84l, MGC1270, P24g1, P24gamma1, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, Tp24, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 11018
Test Specificity Detects human TMED1 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.