Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TMEFF1/Tomoregulin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1919
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1919 500 μg
MAB1919SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1919 Supplier R&D Systems Supplier No. MAB1919
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMEFF1/Tomoregulin-1 Monoclonal specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TMEFF1/Tomoregulin-1
Applications Western Blot
Classification Monoclonal
Clone 249511
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IYR6
Gene Alias C9orf2, H7365member 1, Tomoregulin1, Tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1
Gene Symbols TMEFF1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TMEFF1/Tomoregulin-1 Ser40-Thr328 Accession # Q8IYR6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8577
Test Specificity Detects human TMEFF1/Tomoregulin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TMEFF1 or recombinant human TMEFF2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.