Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TMEFF2/Tomoregulin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1867SP
This item is not returnable.
View return policy
Description
TMEFF2/Tomoregulin-2 Polyclonal specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for Western Blot.Specifications
TMEFF2/Tomoregulin-2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TENB2, Tomoregulin2, Tomoregulin-2, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2 | |
Goat | |
Affinity Purified | |
RUO | |
23671 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q9UIK5 | |
TMEFF2 | |
Mouse myeloma cell line NS0-derived recombinant human TMEFF2/Tomoregulin-2 Arg34-Val320 Accession # Q9UIK5 | |
25 μg | |
Primary | |
Detects human TMEFF2/Tomoregulin-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human TMEFF1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction