Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TMEM119 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB103131T100UG
This item is not returnable.
View return policy
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow CytometrySpecifications
TMEM119 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
OBIF, Osteoblast Induction Factor, TMEM119 | |
Chinese Hamster Ovary cell line CHO derived human TREM119, Arg26-Met96, Accession # Q4V9L6.1 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2699A | |
Flow Cytometry 0.25 ug/10^6 cells | |
Q4V9L6.1 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
338773 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction