Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TMEM59 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9118
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9118 100 μg
MAB9118SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9118 Supplier R&D Systems Supplier No. MAB9118
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMEM59 Monoclonal specifically detects TMEM59 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen TMEM59
Applications Immunocytochemistry
Classification Monoclonal
Clone 816004
Conjugate Unconjugated
Dilution Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BXS4
Gene Alias C1orf8, chromosome 1 open reading frame 8, DCF1, FLJ42977, HSPC001, Liver membrane-bound protein, PRO195, transmembrane protein 59, UNQ169
Gene Symbols TMEM59
Host Species Mouse
Immunogen E.coli-derived recombinant human TMEM59 Met1-Asn220 Accession # Q9BXS4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9528
Test Specificity Detects human TMEM59 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.