Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TMEM87A Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  R&D Systems FAB7966T100UG

Catalog No. FAB7966T100


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

TMEM87A Monoclonal specifically detects TMEM87A in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TMEM87A
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8NBN3
TMEM87A
E. coli-derived recombinant human TMEM87A Ser22-Asn157 Accession # Q8NBN3
RUO
25963
Human
IgG2b
Flow Cytometry
772807
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
DKFZp564G2022, FLJ11273, MGC33727, PSEC0094, transmembrane protein 87A
Mouse
100 μg
Primary
Detects human TMEM87A in ELISAs and Western Blots
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only (RUO)