Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TMIGD2/CD28H Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA83162V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA83162V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA83162V100 Supplier R&D Systems Supplier No. FAB83162V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TMIGD2
Applications Flow Cytometry
Classification Monoclonal
Clone 953743
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q96BF3
Gene Alias CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK-293-derived human TMIGD2/CD28H Met1-Gly150 Accession # Q96BF3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126259
Test Specificity Detects human TMIGD2/CD28H in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.