Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TMIGD2/CD28H Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8316T100UG
This item is not returnable.
View return policy
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry.Specifications
TMIGD2 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
HEK293 human embryonic kidney cell line-derived human TMIGD2/CD28H Met1-Gly150 Accession # Q96BF3 | |
RUO | |
126259 | |
Human | |
IgG2b |
Flow Cytometry | |
953728 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q96BF3 | |
Mouse | |
100 μg | |
Primary | |
Detects human TMIGD2/CD28H in direct ELISAs. Stains human TMIGD2/CD28H transfectants but not irrelevant transfectants in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction