Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TMIGD2/CD28H Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $420.50
Specifications
Antigen | TMIGD2 |
---|---|
Clone | 953728 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8316SP
![]() |
R&D Systems
MAB8316SP |
25 μg |
Each for $214.00
|
|
|||||
MAB8316100
![]() |
R&D Systems
MAB8316100 |
100 μg |
Each for $420.50
|
|
|||||
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
TMIGD2 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
HEK293 human embryonic kidney cell line-derived human TMIGD2/CD28H Met1-Gly150 Accession # Q96BF3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TMIGD2/CD28H in direct ELISAs. Stains human TMIGD2/CD28H transfectants but not irrelevant transfectants in flow cytometry. |
953728 | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q96BF3 | |
126259 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title