Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TMIGD2/CD28H PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB83162P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB83162P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB83162P100 Supplier R&D Systems Supplier No. FAB83162P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TMIGD2
Applications Flow Cytometry
Classification Monoclonal
Clone 953743
Conjugate R-PE
Dilution Flow Cytometry 10 μL/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q96BF3
Gene Alias CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK-293-derived human TMIGD2/CD28H, Accession No. Q96BF3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 126259.0
Test Specificity Detects human TMIGD2/CD28H in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.