Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TMPRSS2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.00
Specifications
Antigen | TMPRSS2 |
---|---|
Clone | 1038105 |
Dilution | Flow Cytometry 0.25 ug/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB107231SP
![]() |
R&D Systems
MAB107231SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10723110
![]() |
R&D Systems
MAB107231100 |
100 μg |
Each for $409.00
|
|
|||||
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Flow CytometrySpecifications
TMPRSS2 | |
Flow Cytometry 0.25 ug/106 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
EC 3.4.21, Epitheliasin, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
Human TMPRSS2 synthetic peptide, Accession # O15393 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1038105 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
O15393 | |
7113 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title