Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB610100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB610100 100 μg
MAB610SP 25 μg
MAB610500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB610100 Supplier R&D Systems Supplier No. MAB610100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 43 publications

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TNF-alpha
Applications Western Blot, Neutralization, Immunocytochemistry, ELISA
Classification Monoclonal
Clone 28401
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.01-0.04 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01375
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Mouse
Immunogen E. coli-derived recombinant human TNF-alpha Gly57-Leu233 (predicted) Accession # P01375
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects human TNF-alpha in ELISAs and Western blots. In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant human TNF-beta , recombinant mouse TNF-alpha , recombinant rat TNF-alpha , and recombinant porcine TNF-alpha is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.